Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MNQLGGLFVN GRPLPLDTRQ QIVRLAVSGM RPCDISRILK VSNGCVSKIL GRYYRTGVLE PKGIGGSKPR LATPPVVARI AQLKGECPAL FAWEIQRQLC AEGLCTQDKT PSVSSINRVL RALQEDQGLP CTRLRSPAVL APAVLTPHSG SETPRGTHPG TGHRNRTIFS PSQAEALEKE FQRGQYPDSV ARGKLATATS LPEDTVRVWF SNRRAKWRRQ EKLKWEMQLP GASQGLTVPR VAPGIISAQQ SPGSVPTAAL PALEPLGPSC YQLCWATAPE RCLSDTPPKA CLKPCWGHLP PQPNSLDSGL LCLPCPSSHC PLASLSGSQA LLWPGCPLLY GLE
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The paired box 4 gene is involved in pancreatic islet development and mouse studies have demonstrated a role for this gene in differentiation of insulin-producing beta cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC129960; paired box gene 4; paired domain gene 4
Gene Aliases: KPD; MODY9
Entrez Gene ID: (Human) 5078
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support