Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 9
PBDC1 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2610029G23Rik; MGC874; Polysaccharide biosynthesis domain-containing protein 1; Protein PBDC1; UPF0368 protein Cxorf26
Gene Aliases: CXorf26; PBDC1
UniProt ID: (Human) Q9BVG4
Entrez Gene ID: (Human) 51260
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support