Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
FIGURE: 1 / 10
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For ICC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE cell sections. For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Involved in transcriptional activation and repression of select genes by chromatin remodeling (alteration of DNA-nucleosome topology).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2310032M22Rik; BAF180; BRG1-associated factor 180; hPB1; Polybromo-1D; Protein polybromo-1
Gene Aliases: 2610016F04Rik; AI507524; BAF180; PB1; PBRM1
UniProt ID: (Human) Q86U86
Entrez Gene ID: (Human) 55193, (Mouse) 66923
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support