Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to PCCA.
Immunogen sequence: LVQEMIRVA KGYPLRHKQA DIRINGWAVE CRVYAEDPYK SFGLPSIGRL SQYQEPLHLP GVRVDSGIQP GSDISIYYDP MISKLITYGS DRTEALKRMA DALDNYVIRG VTHNIALLRE VIINSRFVKG DISTKFLSDV YPDGFKGHML TKSEKNQLLA IASSLFVAFQ LRAQHFQENS RMPVIKPDIA NWELSVKLHD KVHTVVASNN GSVFSVEVDG SKLNVTSTWN LASPLLSVSV DGTQRTVQCL SREAGGNMSI QFLGTVYKVN ILTRLAAELN KFMLEKVTED TSSVLRSPMP GVVVAVSVKP GDAVAEGQEI CVIEAMKMQN SMTAGKTGTV KSVHCQAGDT VGEGDLLVEL E (344-703 aa encoded by BC000140)
This protein is a key enzyme in the catabolic pathway of odd-chain fatty acids, isoleucine, threonine, methionine, and valine. It is probably a dodecamer composed of six biotin-containing alpha subunits and six beta subunits. The protein is located in the mitochondrial matrix. Defects in PCCA are the cause o propionic acidemia type I (PA-1). PA-1 is a life-threatening disease characterized by episodic vomiting, lethargy and ketosis, neutropenia, periodic thrombocytopenia, hypogammaglobulinemia, develop-mental retardation, and intolerance to protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: pccA complementation group; PCCase alpha subunit; PCCase subunit alpha; propanoyl-CoA:carbon dioxide ligase alpha subunit; Propanoyl-CoA:carbon dioxide ligase subunit alpha; propionyl CoA carboxylase, alpha polypeptide; propionyl CoA-carboxylase alpha; Propionyl Coenzyme A carboxylase alpha polypeptide; propionyl Coenzyme A carboxylase, alpha polypeptide; Propionyl-CoA carboxylase alpha chain, mitochondrial; propionyl-coenzyme A carboxylase, alpha polypeptide; RP11-151A6.1
Gene Aliases: C79630; PCCA
UniProt ID: (Human) P05165, (Rat) P14882, (Mouse) Q91ZA3
Entrez Gene ID: (Human) 5095, (Rat) 687008, (Mouse) 110821
Molecular Function:
ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support