Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: ISKLITYGSD RTEALKRMAD ALDNYVIRGV THNIALLREV IINSRFVKGD ISTKFLSDVY PDGFKGHMLT KSEKNQLLAI ASSLFVAFQL RAQHFQENSR MPVIKPDIAN WELSVKLHDK VHTVVASNNG SVFSVEVDGS KLNVTSTWNL ASPLLSVSVD GTQRTVQCLS REAGGNMSIQ FLGTVYKVNI LTRLAAELNK FMLEKVTEDT SSVLRSPMPG VVVAVSVKPG DAVAEGQEIC VIEAMKMQNS MTAGKTGTVK SVHCQAGDTV GEGDLLVELE; Positive Samples: Mouse pancreas, Mouse liver, Mouse brain, Mouse kidney, Rat kidney; Cellular Location: Mitochondrion matrix
This protein is a key enzyme in the catabolic pathway of odd-chain fatty acids, isoleucine, threonine, methionine, and valine. It is probably a dodecamer composed of six biotin-containing alpha subunits and six beta subunits. The protein is located in the mitochondrial matrix. Defects in PCCA are the cause o propionic acidemia type I (PA-1). PA-1 is a life-threatening disease characterized by episodic vomiting, lethargy and ketosis, neutropenia, periodic thrombocytopenia, hypogammaglobulinemia, develop-mental retardation, and intolerance to protein.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PCCase alpha subunit; PCCase subunit alpha; propanoyl-CoA:carbon dioxide ligase alpha subunit; Propanoyl-CoA:carbon dioxide ligase subunit alpha; propionyl CoA-carboxylase alpha; Propionyl Coenzyme A carboxylase alpha polypeptide; Propionyl-CoA carboxylase alpha chain, mitochondrial; propionyl-coenzyme A carboxylase, alpha polypeptide; RP11-151A6.1
Gene Aliases: C79630; Pcca
UniProt ID: (Rat) P14882, (Mouse) Q91ZA3
Entrez Gene ID: (Rat) 687008, (Mouse) 110821
Molecular Function:
ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support