Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
This protein is a key enzyme in the catabolic pathway of odd-chain fatty acids, isoleucine, threonine, methionine, and valine. It is probably a dodecamer composed of six biotin-containing alpha subunits and six beta subunits. The protein is located in the mitochondrial matrix. Defects in PCCA are the cause o propionic acidemia type I (PA-1). PA-1 is a life-threatening disease characterized by episodic vomiting, lethargy and ketosis, neutropenia, periodic thrombocytopenia, hypogammaglobulinemia, develop-mental retardation, and intolerance to protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: pccA complementation group; PCCase alpha subunit; PCCase subunit alpha; propanoyl-CoA:carbon dioxide ligase alpha subunit; Propanoyl-CoA:carbon dioxide ligase subunit alpha; propionyl CoA carboxylase, alpha polypeptide; propionyl Coenzyme A carboxylase, alpha polypeptide; Propionyl-CoA carboxylase alpha chain, mitochondrial; RP11-151A6.1
Gene Aliases: PCCA
UniProt ID: (Human) P05165
Entrez Gene ID: (Human) 5095
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support