Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cadherin-related neuronal receptor c1; PCDH-alpha-C1; protocadherin alpha C1; Protocadherin alpha-C1
Gene Aliases: CNRc1; PCDH-ALPHA-C1; PCDHAC1; rCNRvc1
UniProt ID: (Human) Q9H158
Entrez Gene ID: (Human) 56135, (Mouse) 353236, (Rat) 393091
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support