Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: GSGFGRYSVT EETEKGSFVV NLAKDLGLAE GELAARGTRV VSDDNKQYLL LDSHTGNLLT NEKLDREKLC GPKEPCMLYF QILMDDPFQI YRAELRVRD
Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PCDH-beta-10; protocadherin beta 10; Protocadherin beta-10
Gene Aliases: PCDH-BETA10; PCDHB10; Pcdhb5D; PcdhbJ; PCHB10; UNQ1906/PRO4352
UniProt ID: (Human) Q9UN67
Entrez Gene ID: (Human) 56126, (Mouse) 93881
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support