Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: RHFYYSNRCP KCNIVVHQTQ PLYNIRLDRQ LQDIVYKLVI NLEEREKKQM HDFYKERGLE VPKPAVPQPV PSSKGRSKKV LESVFRIPPE LDMSLLLEFI GANEGTGHFK PLEKKFVRVS GEATIGHVEK FLRRKMGLDP ACQVDIICGD HLLEQYQTLR EIRRAIGDAA MQDGLLVLHY GLVVSPLKIT; Positive Samples: MCF7; Cellular Location: Nucleus
Polycomb group (PcG) proteins form multiprotein complexes that regulate expression patterns of developmental and cell proliferation genes. Several members of the PcG contain ring finger domains and are identified as a subclass of RING finger proteins. The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain the RING-type zinc finger conserved domain are generally involved in the ubiquitination pathway of protein degradation. PCGF6 (polycomb group ring finger 6), also known as MBLR or RNF134, is a 350 amino acid nuclear protein that is ubiquitously expressed and contains one RING-type zinc finger. PCGF6 acts as a transcriptional repressor and regulates the level of histone H3K4Me3 by activating SmcY histone demethylase. PCGF6 exists as two alternatively spliced isoforms.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mel18 and Bmi1-like RING finger; Mel18 and Bmi1-like RING finger protein; Mel18- and Bmi1-like RING finger protein; Polycomb group RING finger protein 6; RING finger protein 134
Gene Aliases: 4933407A11Rik; AI604840; MBLR; PCGF6; RNF134
UniProt ID: (Human) Q9BYE7, (Rat) Q5XI70, (Mouse) Q99NA9
Entrez Gene ID: (Human) 84108, (Rat) 309457, (Mouse) 71041
Molecular Function: histone modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support