Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Positive test controls include: HepG2, Mouse liver, Mouse kidney, Rat liver, Rat pancreas, Rat kidney. The target is usually found in the following locations: Cytoplasm.
Immunogen sequence: FGKYLAHWLS MAQHPAAKLP KIFHVNWFRK DKEGKFLWPG FGENSRVLEW MFNRIDGKAS TKLTPIGYIP KEDALNLKGL GHINMMELFS ISKEFWEKEV EDIEKYLEDQ VNADLPCEIE REILALKQRI SQM
Phosphoenolpyruvate Carboxykinase 1 (PCK1) is a main control point for the regulation of gluconeogenesis. During gluconeogenesis PCK1 acts as the rate-limiting enzyme. PCK1 regulates the formation and maintenance of memory CH8(+) T-cells via gluconeogenesis. PCK1 aids in controlling the levels of metabolic intermediates in the citric acid cycle by regulating cataplerosis and anaplerosis. When glucose levels are high, PCK1 catalyzes the cataplerotic conversion of oxaloacetate to phosphoenolpyruvate (PEP). When they are low it catalyzes the anaplerotic conversion of phosphoenolpyruvate to oxaloacetate. When phosphorylated at Ser-90 by AKT1, PCK1 acts as a protein kinase by reducing the binding affinity to oxaloacetate and promotes an atypical serine protein kinase activity using GTP as the donor. This activity regulates lipogenesis by disrupting the interaction between INSIG proteins and SCAP and promoting nuclear translocation of SREBP proteins and transcription of lipogenesis-related genes. An important paralog of this gene is PCK2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGNC:8724; MGC22652; PE PEPCKC; PEP carboxykinase; PEPCK-C; phosphoenolpyruvate carboxykinase (GTP); phosphoenolpyruvate carboxykinase 1 (soluble); phosphoenolpyruvate carboxykinase 1, cytosolic; phosphoenolpyruvate carboxykinase, cytosolic; phosphoenolpyruvate carboxykinase, cytosolic (GTP); Phosphoenolpyruvate carboxykinase, cytosolic [GTP]; phosphoenolpyruvate carboxylase; phosphopyruvate carboxylase; Serine-protein kinase PCK1
Gene Aliases: AI265463; GTP; PCK; Pck-1; PCK1; PEPCK; PEPCK-C; PEPCK1; PEPCKC; RATPEPCK
UniProt ID: (Human) P35558, (Rat) P07379, (Mouse) Q9Z2V4
Entrez Gene ID: (Human) 5105, (Rat) 362282, (Mouse) 18534
Molecular Function:
kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support