Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: GGMYHNTFTI QCSLPVFLSI SVVDQPDLDP QFVREFYSAS VAEDAAKGTS VLTVEAVDGD KGINDPVIYS ISYSTRPGWF DIGADGVIRV NGSLDREQLL EADEEVQLQ
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cadherin; Cadherin-related family member 2; PC-LKC; protocadherin 24; Protocadherin LKC; Protocadherin-24
Gene Aliases: CDHR2; PCDH24; PCLKC
UniProt ID: (Human) Q9BYE9
Entrez Gene ID: (Human) 54825
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support