Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
1 µg/mL of MA5-45258 was sufficient for detection of Piccolo in 20 µg of rat brain tissue lysate by colorimetric immunoblot analysis using goat IgG:HRP as the secondary antibody.|Detects approximately 550kDa. No cross-reactivity to Bassoon.
The protein encoded by the PCLO gene is part of the presynaptic cytoskeletal matrix, which is involved in establishing active synaptic zones and in synaptic vesicle trafficking. Variations in this gene have been associated with bipolar disorder and major depressive disorder. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aczonin; Brain-derived HLMN protein; Multidomain presynaptic cytomatrix protein; multidomain presynaptic cytomatrix protein Piccolo; PCLO; presynaptic cytomatrix protein; Protein piccolo
Gene Aliases: ACZ; aczonin; KIAA0559; PCH3; PCLO; Pico
UniProt ID: (Human) Q9Y6V0, (Mouse) Q9QYX7, (Rat) Q9JKS6
Entrez Gene ID: (Human) 27445, (Mouse) 26875, (Rat) 56768
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support