Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
PCNP is a protein coding gene, and may be involved in cell cycle regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PCNP; PEST proteolytic signal-containing nuclear protein; PEST-containing nuclear protein
Gene Aliases: PCNP
UniProt ID: (Human) Q8WW12
Entrez Gene ID: (Human) 57092
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support