Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen Sequence: EKTEESPSAP DAPTCPKQCR RTGTLQSNFC ASSLVVTATV KSMVREPGEG LAVTVSLIGA YKTGGLDLPS PPTGASLKFY VPCKQCPPMK KGVSYLLMGQ VEENRGPVLP PESFVVLHRP NQDQILTNLS KRKCPSQPVR AAASQD
Fibrillar collagen types I-III are synthesized as precursor molecules known as procollagens. These precursors contain amino- and carboxyl-terminal peptide extensions known as N- and C-propeptides, respectively, which are cleaved, upon secretion of procollagen from the cell, to yield the mature triple helical, highly structured fibrils. This gene encodes a glycoprotein which binds and drives the enzymatic cleavage of type I procollagen and heightens C-proteinase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: astroglial cell transcript 2; P14; PCPE-1; Procollagen C-endopeptidase enhancer 1; procollagen C-endopeptidase enhancer protein; procollagen C-proteinase enhancer 1; procollagen C-proteinase enhancer protein; Procollagen COOH-terminal proteinase enhancer 1; procollagen, type 1, COOH-terminal proteinase enhancer; Type 1 procollagen C-proteinase enhancer protein; Type I procollagen COOH-terminal proteinase enhancer
Gene Aliases: Astt-2; Astt2; P14; PCOLCE; PCPE; PCPE-1; PCPE1
UniProt ID: (Human) Q15113, (Mouse) Q61398, (Rat) O08628
Entrez Gene ID: (Human) 5118, (Mouse) 18542, (Rat) 29569
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support