Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: WVEDVLDKF MRIYRYQSHD YAFSSVEKLL HALGGDDFLG MLNRTLLETL QKAGFSEKFL NEMIAPVMRV NYGQSTDINA FVGAVSLSCS DSGLWAVEGG NKLVCSGLLQ ASKSNLISGS VMYIEEKTKT KYTGNPTKMY EVVYQIGTET RSDFYDIVLV ATPLNRKMSN ITFLNFDPPI EEFHQYYQHI VTTLVKGELN TSIFSSRPID KFGLNTVLTT DNSDLFINSI GIVPSVREKE DPEPSTDGTY VWKIFSQETL TKAQILKLFL SYDYAVKKPW LAYPHYKPPE KCPSIILHDR LYYLNGIECA ASAMEMSAIA AHNAALLAYH RWNGHTDMID QDGLYEKLKT EL (155-505 aa encoded by BC033815)
Prenylcysteine is released during the degradation of prenylated proteins. PCYOX1 catalyzes the degradation of prenylcysteine to yield free cysteines and a hydrophobic isoprenoid product (Tschantz et al., 1999 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chloride ion pump-associated 55 kDa protein; Prenylcysteine lyase; prenylcysteine oxidase; Prenylcysteine oxidase 1
Gene Aliases: 1200015P13Rik; AI115532; AI448340; Clp55; KIAA0908; PCL1; Pcly; PCYOX1; UNQ597/PRO1183
UniProt ID: (Human) Q9UHG3, (Mouse) Q9CQF9, (Rat) Q99ML5
Entrez Gene ID: (Human) 51449, (Mouse) 66881, (Rat) 246302
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support