Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/ml.
Programmed cell death 1 ligand 1 is one of the two ligands of PD-1. PD-L1 is expressed on macrophages, T cells, B cells, NK cells, DCs and some cancer cell surface. Binding of PD-1 with PD-L1 could result in down-regulation of the immune system by inhibiting the T-cell activation process. Thus, PD-L1 is an important immune checkpoint and popular target for therapeutic antibodies against many cancers. GenScript Human PD-L1 Antibody (PDL1.D1), mAb, Mouse is produced from the hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from mouse immunized with recombinant human PD-L1-Fc(Z03371).
Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B7 homolog 1; B7-H1; CD274; CD274 antigen; PD-L1; PDCD1 ligand 1; pdl-1; Programmed cell death 1 ligand 1; sCD274; soluble CD274; soluble PD L1; sPD L1
Gene Aliases: B7-H; B7H1; CD274; PD-L1; PDCD1L1; PDCD1LG1; PDL1
UniProt ID: (Human) Q66RK1
Entrez Gene ID: (Human) 29126
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support