Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LLTRHNLISR FKIPTVFLMS FLDALETGYG KYKNPYHNQI HAADVTQTVH CFLLRTGMVH CLSEIELLAI IFAAAIHDYE HTGTTNSFHI QTKSECAIVY NDRSVLENHH ISSVFRLMQD DEMNIFINLT KDEFVELRAL VIEMVLATDM SCHFQQVKTM KTALQQLERI DKPKALSLLL HAADISHPTK QWLVHSRWTK ALMEEFFRQG DKEAELGLPF SPLCDRTSTL VAQSQIGFID FIVEPTFSVL TDVAEKSVQP LADEDSKSKN QPSFQWRQPS LDVEVGDPNP DVVSFRSTWV KRIQENKQKW KERAASGITN QMSIDELSPC EEEAPPSPAE DEHNQNGNLD
The protein encoded by this gene belongs to the cyclic nucleotide phosphodiesterase (PDE) family, and PDE1 subfamily. Members of the PDE1 family are calmodulin-dependent PDEs that are stimulated by a calcium-calmodulin complex. This PDE has dual-specificity for the second messengers, cAMP and cGMP, with a preference for cGMP as a substrate. cAMP and cGMP function as key regulators of many important physiological processes. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 63 a Cam PDE; 63 a Cam-PDE; 63 kDa Cam-PDE; anti-oxidant protein 2; calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1B; calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase; calmodulin-stimulated phosphodiesterase PDE1B1; Cam PDE 1B; Cam PDE1B; Cam-PDE 1B; cyclic nucleotide phosphodiesterase (CaM-PDE); Dual specificity calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1B; epididymis secretory sperm binding protein Li 79p; PDE 1B; PDES 1B; Phosphodiesterase; phosphodiesterase 1B, Ca2+-calmodulin dependent, 63 kDa; phosphodiesterase 1B, Ca2+calmodulin dependent; phosphodiesterase 1B, calmodulin-dependent; phosphodiesterase 1B1 Ca2+-calmodulin dependent 63 kDa; phosphodiesterase 1B1, Ca2+-calmodulin dependent, 63 kDa; phosphodiesterase IB calmodulin-dependent; presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase family known as PDE1B
Gene Aliases: Aop2; HEL-S-79p; PDE1B; PDE1B1; PDES1B
UniProt ID: (Human) Q01064, (Mouse) Q01065, (Rat) Q01066
Entrez Gene ID: (Human) 5153, (Mouse) 18574, (Rat) 29691
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support