Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: PLTPFPGFYP CSEIEDPAEK GDRKLNKGLN RNSLPTPQLR RSSGTSGLLP VEQSSRWDRN NGKRPHQEFG ISSQGCYLNG PFNSNLLTIP KQRSSSVSLT HHVGLRRAGV LSSLSPVNSS NHGPVSTGSL TNRSPIEFPD TADFLNKPSV ILQRSLGNAP NTPDFYQQLR NSDSNLCNSC GHQMLKYVST SESDGTDCCS GKSGEEENIF SKESFKLMET QQEEETEKKD SRKLFQEGDK WLTEEAQSEQ QTNIEQEVSL D
Cyclic nucleotide phosphodiesterase with a dual-specificity for the second messengers cAMP and cGMP, which are key regulators of many important physiological processes. May play a role in fat metabolism. Regulates cAMP binding of RAPGEF3. Through simultaneous binding to RAPGEF3 and PIK3R6 assembles a signaling complex in which the PI3K gamma complex is activated by RAPGEF3 and which is involved in angiogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CGI-PDE B; CGIP1; CGIPDE1; cGMP-inhibited 3',5'-cyclic phosphodiesterase 3B; cGMP-inhibited 3',5'-cyclic phosphodiesterase B; Cyclic GMP-inhibited phosphodiesterase B; cyclic nucleotide phosphodiesterase; phosphodiesterase 3B, cGMP-inhibited
Gene Aliases: 9830102A01Rik; AI847709; cGIPDE1; HcGIP1; PDE3B
UniProt ID: (Human) Q13370, (Mouse) Q61409
Entrez Gene ID: (Human) 5140, (Mouse) 18576
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support