Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVIDMVLAT DMSKHMTLLA DLKTMVETKK VTSSGVLLLD NYSDRIQVLR NMVHCADLSN PTKPLELYRQ WTDRIMAEFF QQGDRERERG MEISPMCDKH TASVEKSQVG FIDYIVHPLW ETWADLVHPD AQEILDTLED NRDWYYSAIR QSPSPPPEEE SRGPGHPPLP DKFQFELTLE EEEEEEISMA QIPCTAQEAL TAQGLSGVEE ALDATIAWEA SPAQESLEVM AQEASLEAEL EAVYLTQQAQ STGSAPVAPD EFSSREEFVV AVSHSSPSAL ALQSPLLPAW RTLSVSEHAP GLPGLPSTAA EVEAQREHQA AKRACSACAG TFGEDTSALP APGGGGSGGD PT (297-647 aa encoded by BC019864)
Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. Cyclic nucleotide phosphodiesterases (PDEs) regulate the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. PDE4A is a class IV cAMP-specific PDE.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3',5'-cyclic-AMP phosphodiesterase 4A; cAMP-specific 3',5'-cyclic phosphodiesterase 4A; cAMP-specific phosphodiesterase 4A; cAMP-specific phosphodiesterase PDE4A; cyclic AMP specific phosphodiesterase PDE4A5A; DPDE2; dunce; PDE46; phosphodiesterase 4A, cAMP specific; phosphodiesterase 4A, cAMP-specific (dunce; phosphodiesterase 4A, cAMP-specific (phosphodiesterase E2 dunce homolog, Drosophila); phosphodiesterase E2 dunce homolog, Drosophila; phosphodiesterase isozyme 4
Gene Aliases: D9Ertd60e; DPDE2; PDE4; PDE46; PDE4A; PHOSA
UniProt ID: (Human) P27815, (Mouse) O89084, (Rat) P54748
Entrez Gene ID: (Human) 5141, (Mouse) 18577, (Rat) 25638
Molecular Function:
phosphodiesterase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support