Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ETSLLENKR NQVLLDLASL IFEEQQSLEV ILKKIAATII SFMQVQKCTI FIVDEDCSDS FSSVFHMECE ELEKSSDTLT REHDANKINY MYAQYVKNTM EPLNIPDVSK DKRFPWTTEN TGNVNQQCIR SLLCTPIKNG KKNKVIGVCQ LVNKMEENTG KVK (321-482 aa encoded by BC126233)
The cyclic monophosphate nucleotides (cyclic adenosine monophosphate [cAMP] and cyclic guanosine monophosphate [cGMP] are found ubiquitously in mammalian cells and act as second messenger transducers to effect the intracellular actions of a variety of G protein coupled receptors (GPCRs) for hormones, cytokines, and neurotransmitters. Cyclic nucleotides are important intracellular second messenger which play important role in a variety of signal transduction process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CGB-PDE; cGMP-binding cGMP-specific 3',5'-cyclic nucleotide phosphodiesterase; cGMP-binding cGMP-specific phosphodiesterase; cGMP-binding/cGMP-specific phosphodiesterase; cGMP-specific 3',5'-cyclic phosphodiesterase; cGMP-specific phosphodiesterase PDE5A2; cGMP-specific phosphodiesterase type 5A; phosphodiesterase 5A, cGMP-specific; phosphodiesterase isozyme 5
Gene Aliases: CGB-PDE; Cgbpde; CN5A; Cn5n; PDE5; PDE5A; Pde5a1
UniProt ID: (Human) O76074, (Mouse) Q8CG03
Entrez Gene ID: (Human) 8654, (Mouse) 242202
Molecular Function: phosphodiesterase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support