Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAASWRLGC DPRLLRYLVG FPGRRSVGLV KGALGWSVSR GANWRWFHST QWLRGDPIKI LMPSLSPTME EGNIVKWLKK EGEAVSAGDA LCEIETDKAV VTLDASDDGI LAKIVVEEGS KNIRLGSLIG LIVEEGEDWK HVEIPKDVGP PPPVSKPSEP RPSPEPQISI PVKKEHIPGT LRFRLSPAAR NILEKHSLDA SQGTATGPRG IFTKEDALKL VQLKQTGKIT ESRPTPAPTA TPTAPSPLQA TAGPSYPRPV IPPVSTPGQP NAVGTFTEIP ASNIRRVIAK RLTESKSTVP HAYATADCDL GAVLKVRQDL VKDDIKVSVN DFIIKAAAVT LKQMPDVNVS WDGEGPKQLP FIDISVAVAT VKGLLTPIIK DAAAKGIQEI ADSVKALSKK ARDGKLLPEE YQGGSFSISN LGMFGIDEFT AVINPPQACI LAVGRFRPVL KLTEDEEGNA KLQQRQLITV TMSSDSRVVD DELATRFLKS FKANLENPIR LA (1-501 aa encoded by BC010389)
The PDHX gene encodes component X of the pyruvate dehydrogenase (PDH) complex. For a detailed description of the pyruvate dehydrogenase complex. The mammalian PDH complex differs from that in E. coli and from the other mammalian alpha-keto acid dehydrogenases by the presence of a 53-kDa protein called protein X. Component X binds to the E3 component of the PDH complex (Robinson et al., 1990 ; Aral et al., 1997 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: component X; Dihydrolipoamide dehydrogenase-binding protein of pyruvate dehydrogenase complex; dihydrolipoamide dehydrogenase-binding protein of pyruvate dehydrogenase complex (E3-binding protein) (proX); E3-binding protein; E3BP; IPF1; lipoyl-containin; Lipoyl-containing pyruvate dehydrogenase complex component X; proX; pyruvate dehydrogenase; pyruvate dehydrogenase complex, component X; pyruvate dehydrogenase complex, E3-binding protein subunit; pyruvate dehydrogenase complex, lipoyl-containing component X; Pyruvate dehydrogenase protein X component, mitochondrial
Gene Aliases: Ac1164; AI481367; DLDBP; E3BP; OPDX; PDHX; PDX1; proX
UniProt ID: (Human) O00330, (Mouse) Q8BKZ9
Entrez Gene ID: (Human) 8050, (Rat) 311254, (Mouse) 27402
Molecular Function:
acetyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support