Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Immunogen sequence: FHEKSPDDQK ALSDFVDTLI KVRNRHHNVV PTMAQGIIEY KDACTVDPVT NQNLQYFLDR FYMNRISTRM LMNQHILIFS DSQTGNPSHI GSIDPNCDVV AVVQDAFECS RMLCDQYYLS SPELKLTQVN GKFPDQPIHI VYVPSHLHHM LFELFKNAMR ATVEHQENQP SLTPIEVIVV LGKEDLTIKI SDRGGGVPLR IIDRLFSYTY STAPTPVMDN SRNAPLAGFG YGLPISRLYA KYFQGDLNLY SLSGYGTDAI IYLKALSSES IEKLPVFNKS AFKHYQMSSE ADDWCIPSRE PKNLAKEVAM
PDK4 (PDHK4), along with the other PDHKs, regulates glucose metabolism by phosphorylating the E1 alpha subunit of the mitochondrial pyruvate dehydrogenase complex. Inhibition of PDHKs is viewed as a potential therapeutic treatment for type II diabetes. This protein is located in the matrix of the mitrochondria and it's expression is regulated by glucocorticoids, retinoic acid and insulin. PDK4 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 4, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 4, mitochondrial; kinase isozyme 4; lipoamide; pyruvate dehydrogenase; pyruvate dehydrogenase kinase 4; Pyruvate dehydrogenase kinase isoform 4; pyruvate dehydrogenase kinase, isoenzyme 4; pyruvate dehydrogenase kinase, isozyme 4; pyruvate dehydrogenase, lipoamide, kinase isozyme 4, mitochondrial; pyruvate dehydrogenate kinase 4
Gene Aliases: AV005916; PDHK4; PDK4
UniProt ID: (Human) Q16654, (Rat) O54937, (Mouse) O70571
Entrez Gene ID: (Human) 5166, (Rat) 89813, (Mouse) 27273
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support