Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Nop56p is a yeast nucleolar protein that is part of a complex with the nucleolar proteins Nop58p and fibrillarin. Nop56p is required for assembly of the 60S ribosomal subunit and is involved in pre-rRNA processing. NOL5A is similar in sequence to Nop56p and is also found in the nucleolus. Multiple transcript variants encoding several different isoforms have been found for this gene, but the full-length nature of most of them have not been determined.Nop56p is a yeast nucleolar protein that is part of a complex with the nucleolar proteins Nop58p and fibrillarin. Nop56p is required for assembly of the 60S ribosomal subunit and is involved in pre-rRNA processing. The protein encoded by this gene is similar in sequence to Nop56p and is also found in the nucleolus. Multiple transcript variants encoding several different isoforms have been found for this gene, but the full-length nature of most of them have not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell proliferation-inducing gene 54 protein; PDS5, regulator of cohesion maintenance, homolog A; PDS5A; SCC-112; Sister chromatid cohesion protein 112; Sister chromatid cohesion protein PDS5 homolog A
Gene Aliases: 9030416H16Rik; E230024D05Rik; KIAA0648; mKIAA0648; PDS5; PDS5A; PIG54; RGD1307094; SCC-112; SCC112
UniProt ID: (Human) Q2TTR5, (Rat) A4L9P7
Entrez Gene ID: (Human) 23244, (Rat) 305343, (Mouse) 71521
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support