Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: MDASLEKIAD PTLAEMGKNL KEAVKMLEDS QRRTEEENGK KLISGDIPGP LQGSGQDMVS ILQLVQNLMH GDEDEEPQSP RIQNIGEQGH MALLGHSLGA YISTLDKEKL RKLTTRILSD TTLWLCRIFR YENGCAYFHE EEREGLAKIC RLAIHSRYED FVVDGFNVLY NKKPVIYLSA AARPGLGQYL CNQDVAFLEK LIKDDIERGR LPLLLVANAG TAAVGHTDKI GRLKELCEQY GIWLHVEGVN LATLALGYVS SSVLAAAKCD SMTMTPGPWL GLPAVPAVTL YKHDDPALTL VAGLTSNKPT DKLRALPLWL SLQYLGLDGF VERIKHACQL SQRLQ
PDXDC1 (Pyridoxal Dependent Decarboxylase Domain Containing 1) is a Protein Coding gene. Diseases associated with PDXDC1 include Autosomal Recessive Nonsyndromic Deafness 32 and Fetal Akinesia Deformation Sequence 1. Gene Ontology (GO) annotations related to this gene include pyridoxal phosphate binding and carboxy-lyase activity. An important paralog of this gene is HDC. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: pyridoxal-dependent decarboxylase domain containing 1; Pyridoxal-dependent decarboxylase domain-containing protein 1
Gene Aliases: 2210010A19Rik; AA415817; KIAA0251; Kiaa0251-hp; LP8165; PDXDC1; RGD1562597
UniProt ID: (Human) Q6P996, (Mouse) Q99K01
Entrez Gene ID: (Human) 23042, (Pig) 100622608, (Mouse) 94184, (Rat) 304721
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support