Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: KRYGGFLRRI RPKLKWDNQK RYGGFLRRQF KVVTRSQEDP NAYSGELFDA
expression in hippocampus increases in response to traumatic brain injury; may play a role in regulatio of neurotransmitter release.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-neoendorphin-dynorphin; leu-enkephalin; leumorphin; neoendorphin-dynorphin-enkephalin prepropeptide; Preprodynorphin; preproenkephalin B; Proenkephalin-B; rimorphin
Gene Aliases: ADCA; PDYN; PENKB; SCA23
UniProt ID: (Human) P01213
Entrez Gene ID: (Human) 5173
Molecular Function:
neuropeptide
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support