Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MEKAADLQDT ASLTLKFKFN PKLGIDNPVL SLAEDHDPYD PWSLERPRFC LLSKEEGKSF GFHLQQELGR AGHVVCRVDP GTSAQRQGLQ EGDRILAVNN DVVEHEDYAV VVRRIRASSP RVLLTVLARH AHDVARAQLG EDAHLCPTLG PGVRPRLCHI VKDEGGFGFS VTHGNQGPFW LVLSTGGAAE RAGVPPGARL LEVNGLWQSG QQVTLLVAGP EVEEQCRQLG LPLAAPLAEG WALPTKPRCL HLEKGPQGFG FLLREEKGLD GRPGQFLWEV DPGLPAKKAG MQAGDRLVAV AGESVEGLGH EETVSRIQGQ GSCVSLTVVD PEADRFFSMV RLSPLLFLEN TEAPASPQGS SSASLVETED PSLEDTSVPS VPLGSRQCFL YPGPGGSYGF RLSCVASGPR LFISQVTPGG SAARAGLQVG DVILEVNGYP VGGQNDLERL QQLPEAEPPL CLKLAARSLR GLEAWIPPGA AEDWALASDL L
Acts as a regulatory protein that associates with GUCY2C and negatively modulates its heat-stable enterotoxin-mediated activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: intestinal and kidney enriched PDZ protein; Intestinal and kidney-enriched PDZ protein; Na(+)/H(+) exchange regulatory cofactor NHE-RF4; Na/Pi cotransporter C-terminal-associated protein 2; naPi-Cap2; Natrium-phosphate cotransporter IIa C-terminal-associated protein 2; NHERF-4; PDZ domain containing 2; PDZ domain-containing protein 2; PDZ domain-containing protein 3; Sodium-hydrogen exchanger regulatory factor 4
Gene Aliases: DLNB27; IKEPP; NHERF4; PDZD3; PDZK2
UniProt ID: (Human) Q86UT5
Entrez Gene ID: (Human) 79849
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support