Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: RAFYGLKQVE KANEAMAIDT LLISDELFRH QDVATRSRYV RLVDSVKENA GTVRIFSSLH VSGEQLSQLT GVAAILRFPV PELSDQEGDS SSEED
PELO encodes a protein which contains a conserved nuclear localization signal. The encoded protein may have a role in spermatogenesis, cell cycle control, and in meiotic cell division.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPelota; Protein Dom34 homolog; Protein pelota homolog
Gene Aliases: CGI-17; PELO; PRO1770
UniProt ID: (Human) Q9BRX2
Entrez Gene ID: (Human) 53918
Molecular Function:
translation release factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support