Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAATGPSFW LGNETLKVPL ALFALNRQRL CERLRKNPAV QAGSIVVLQG GEETQRYCTD TGVLFRQESF FHWAFGVTEP GCYGVIDVDT GKSTLFVPRL PASHATWMGK IHSKEHFKEK YAVDDVQYVD EIASVLTSQK PSVLLTLRGV NTDSGSVCRE ASFDGISKFE VNNTILHPEI VECRVFKTDM ELEVLRYTNK ISSEAHREVM KAVKVGMKEY ELESLFEHYC YSRGGMRHSS YTCICGSGEN SAVLHYGHAG APNDRTIQNG DMCLFDMGGE YYCFASDITC SFPANGKFTA DQKAVYEAVL RSSRAVMGAM KPGVWWPDMH RLADRIHLEE LAHMGILSGS VDAMVQAHLG AVFMPHGLGH FLGIDVHDVG GYPEGVERID EPGLRSLRTA RHLQPGMVLT VEPGIYFIDH LLDEALADPA RASFFNREVL QRFRGFGGVR IEEDVVVTDS GIELLTCVPR TVEEIEACMA GCDKAFTPFS GPK
Xaa-Pro dipeptidase is a cytosolic dipeptidase that hydrolyzes dipeptides with proline or hydroxyproline at the carboxy terminus (but not Pro-Pro). It is important in collagen metabolism because of the high levels of iminoacids. Xaa-Pro dipeptidase is a cytosolic dipeptidase that hydrolyzes dipeptides with proline or hydroxyproline at the carboxy terminus (but not Pro-Pro). It is important in collagen metabolism because of the high levels of iminoacids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aminoacyl-L-proline hydrolase; Imidodipeptidase; MGC10905; Peptidase D; Prolidase; Proline dipeptidase; testicular tissue protein Li 138; X-Pro dipeptidase; Xaa-Pro dipeptidase
Gene Aliases: PEPD; PRD; PROLIDASE
UniProt ID: (Human) P12955
Entrez Gene ID: (Human) 5184
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support