Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSGPLEGAD GGGDPRPGES FCPGGVPSPG PPQHRPCPGP SLADDTDANS NGSSGNESNG HESRGASQRS SHSSSSGNGK DSALLETTES SKSTNSQSPS PPSSSIAYSL LSASSEQDNP STSGCSSEQS ARARTQKELM TALRELKLRL PPERRGKGRS GTLATLQYAL ACVKQVQANQ EYYQQWSLEE GEPCSMDMST YTLEELEHIT SEYTLQNQDT FSVAVSFLTG RIVYISEQAA VLLRCKRDVF RGTRFSELLA PQDVGVFYGS TAPSRLPTWG TGASAGSGLR DFTQEKSVFC RIRGGPDRDP GPRYQPFRLT PYVTKIRVSD GAPAQPCCLL IAE (1-341 aa encoded by BC028207)
Circadian rhythmicity is a basic property of phylogenetically diverse organisms which range from animals and plants, to fungi. Regulation of endogenous biological clocks is regulated at the genetic level by a protein-mediated, autoregulatory feed-back loop. In mammals, several genes that encode members of the basic helix-loop helix (bHLH) PAS (PER-ARNT-SIM) transcription factor family have been shown to play a significant role in regulating circadian oscillations. Transactivation of CLOCK-induced genes is mediated via an E box enhancer (CACGTG) found upstream of target genes. CLOCK-ARNT3 heterodimers bind to E box regulatory elements and stimulate gene transcription. CLOCK has been shown to transactivate the mammalian homolog of Drosophila per. PER, in concert with the product of the mammalian timeless gene (TIM), negatively regulates its own transcription by blocking the activity of the CLOCK-BMAL1 transactivation complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Circadian clock protein PERIOD 1; Circadian pacemaker protein Rigui; hPER1; mPER1; per1 {ECO:0000312|RGD:727863}; period 1; Period circadian protein homolog 1; period homolog 1; Period, drosophila, homolog of; rPER1
Gene Aliases: hPER; KIAA0482; m-rigui; mPer1; PER; PER1; RIGUI; rper1
UniProt ID: (Human) O15534, (Mouse) O35973, (Rat) Q8CHI5
Entrez Gene ID: (Human) 5187, (Mouse) 18626, (Rat) 287422
Molecular Function:
transcription cofactor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support