Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MPRGEAPGPG RRGAKDEALG EESGERWSPE FHLQRKLADS SHSEQQDRNR VSEELIMVVQ EMKKYFPSER RNKPSTLDAL NYALRCVHSV QANSEFFQIL SQNGAPQADV SMYSLEELAT IASEHTSKNT DTFVAVFSFL SGRLVHISEQ AALILNRKKD VLASSHFVDL LAPQDMRVFY AHTARAQLPF WNNWTQRAAR YECAPVKPFF CRIRGGEDRK QEKCHSPFRI IPYLIHVHHP AQPELESEPC CLTVVEKIHS GYEAPRIPVN KRIFTTTHTP GCVFLEVDEK AVPLLGYLPQ DLIGTSILSY LHPEDRSLMV AIHQKVLKYA GHPPFEHSPI RFCTQNGDYI ILDSSWSSFV NPWSRKISFI IGRHKVRT
This gene is a member of the Period family of genes and is expressed in a circadian pattern in the suprachiasmatic nucleus, the primary circadian pacemaker in the mammalian brain. Genes in this family encode components of the circadian rhythms of locomotor activity, metabolism, and behavior. This gene is upregulated by CLOCK/ARNTL heterodimers but then represses this upregulation in a feedback loop using PER/CRY heterodimers to interact with CLOCK/ARNTL. Polymorphisms in this gene have been linked to sleep disorders. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell growth-inhibiting gene 13 protein; Circadian clock protein PERIOD 3; hPER3; period circadian protein 3; Period circadian protein homolog 3
Gene Aliases: FASPS3; GIG13; PER3
UniProt ID: (Human) P56645
Entrez Gene ID: (Human) 8863
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support