Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGGLEKKKYE RGSATNYITR NKARKKLQLS LADFRRLCIL KGIYPHEPKH KKKVNKGSTA ARTFYLIKDI RFLLHEPIVN KFREYKVFVR KLRKAYGKSE WNTVERLKDN KPNYKLDHII KERYPTFIDA LRDLDDALSM CFLFSTFPRT GKCHVQTIQL CRRLTVEFMH YIIAARALRK VFLSIKGIYY QAEVLGQPIV WITPYAFSHD HPTDVDYRVM ATFTEFYTTL LGFVNFRLYQ LLNLHYPPKL EGQAQAEAKA GEGTYALDSE SCMEKLAALS ASLARVVVPA TEEEAEVDEF PTDGEMSAQE EDRRKELEAQ EKHKKLFEGL KFFLNREVPR EALAFIIRSF GGEVSWDKSL CIGATYDVTD SRITHQIVDR PGQQTSVIGR CYVQPQWVFD SVNARLLLPV AEYFSGVQLP PHLSPFVTEK EGDYVPPEKL KLLALQRGED PGNLNESEEE EEEDDNNEGD GDEEGENEEE EEDAEAGSEK EEEARLAALE EQRMEGKKPR VMAGTLKLED KQRLAQEEES EAKRLAIMMM KKREKYLYQK IMFGKRRKIR EANKLAEKRK AHDEAVRSEK KAKKARPE
Phosphomannose isomerase catalyzes the interconversion of fructose-6-phosphate and mannose-6-phosphate and plays a critical role in maintaining the supply of D-mannose derivatives, which are required for most glycosylation reactions. Mutations in the MPI gene were found in patients with carbohydrate-deficient glycoprotein syndrome, type Ib.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PES1; Pescadillo homolog; pescadillo homolog 1, containing BRCT domain
Gene Aliases: PES; PES1
UniProt ID: (Human) O00541
Entrez Gene ID: (Human) 23481
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support