Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LLSTHSDEV TDSINWASGE DDHVVARAEY DFAAVSEEEI SFRAGDMLNL ALKEQQPKVR GWLLASLDGQ TTGLIPANYV KILGKRKGRK TVESSKVSKQ QQSFTNPTLT KGATVADSLD EQEAAFESVF VETNKVPVAP DSIGKDGEKQ DL (253-403 aa encoded by BC067090)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Peroxin-13; Peroxisomal membrane protein PEX13
Gene Aliases: NALD; PBD11A; PBD11B; PEX13; ZWS
UniProt ID: (Human) Q92968
Entrez Gene ID: (Human) 5194
Molecular Function:
primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support