Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LESPDFSTVL NTCLNRGFSR LLDNMAEFFR PTEQDLQHGN SMNSLSSVSL PLAKIIPIVN GQIHSVCSET PSHFVQDLLT MEQVKDFAAN VYEAFSTPQQ LEK
Peroxin 3, also known as PEX3 (peroxisomal biogenesis factor 3) or TRG18, is a 373 amino acid multi-pass membrane protein that localizes to peroxisomes and belongs to the Peroxin family. Expressed ubiquitously, Peroxin 3 interacts with Peroxin 19 and is involved in peroxisome biosynthesis and membrane vesicle assembly, as well as in the maintenance of peroxisomal integrity. Additionally, Peroxin 3 acts as a docking factor for Peroxin 19 and is required for the import of peroxisomal proteins. Defects in the gene encoding Peroxin 3 are the cause of peroxisome biogenesis disorder complementation group 12 (PBD-CG12) and Zellweger syndrome (ZwS), both of which arise from a failure of peroxisomal protein import.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Peroxin-3; Peroxin3; Peroxisomal assembly protein PEX3; Peroxisomal biogenesis factor 3; transformation-related protein 18
Gene Aliases: PBD10A; PEX3; TRG18
UniProt ID: (Human) P56589
Entrez Gene ID: (Human) 8504
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support