Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to PGAP1.
Involved in inositol deacylation of GPI-anchored proteins. GPI inositol deacylation may important for efficient transport of GPI-anchored proteins from the endoplasmic reticulum to the Golgi.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GPI inositol-deacylase; hPGAP1; Post-GPI attachment to proteins factor 1
Gene Aliases: 5033403E17Rik; 9030223K07Rik; A530084K22; Bst1; D230012E17Rik; ISPD3024; MRT42; oto; PGAP1; SPG67; UNQ3024/PRO9822
UniProt ID: (Human) Q75T13, (Mouse) Q3UUQ7
Entrez Gene ID: (Human) 80055, (Mouse) 241062
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support