Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YGPSLMPGGN KEAWPHIKTI FQGIAAKVGT GEPCCDWVGD EGAGHFVKMV HNGIEYGDMQ LICEAYHLMK DVLGMAQDEM AQAFEDWNKT ELDSFLIEIT ANILKFQDTD GKHLLPKIRD SAGQKGTGKW TAISALEYGV PVTLIGEAVF ARCLSSLKDE RIQASKKLKG PQKFQFDGDK KSFLEDIRKA LYASKIISYA QGFMLLRQAA TEFGWTLNYG GIALMWRGGC IIRSVFLGKI KDAFDRNPEL QNLLLDDFFK SAVENCQDSW RRAVSTGVQA GIPMPCFTTA LSFYDGYRHE MLPASLIQAQ RDYFGAHTYE LLAKPGQFIH TNWTGHGGTV SSSSYNA
6-phosphogluconate dehydrogenase is the second dehydrogenase in the pentose phosphate shunt. Deficiency of this enzyme is generally asymptomatic, and the inheritance of this disorder is autosomal dominant. Hemolysis results from combined deficiency of 6-phosphogluconate dehydrogenase and 6-phosphogluconolactonase suggesting a synergism of the two enzymopathies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 6-phosphogluconate dehydrogenase, decarboxylating
Gene Aliases: 6PGD; PGD; PGDH
UniProt ID: (Human) P52209
Entrez Gene ID: (Human) 5226
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support