Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YTFLKVLNNM EIGASLFDEE GAKIVKDIMA KAQKNGVRIT FPVDFVTGDK FDENAQVGKA TVASGISPGW MGLDCGPESN KNHAQVVAQA RLIVWNGPLG VFEWDAFAKG TKALMDEIV
PGK2 is a form of phosphoglycerate kinase unique to spermatozoa. The autosomal PGK2 gene lacks introns and contains characteristics of a processed gene. The conservation of function in this processed PGK2 gene and its tissue-specific expression in spermatogenesis suggests that it exists as a compensatory response to the inactivation of the X-linked PGK1 gene in spermatogenic cells before meiosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 272; Phosphoglycerate kinase 2; phosphoglycerate kinase autosomal pseudogene; Phosphoglycerate kinase, testis specific; testicular tissue protein Li 139; testis-specific phosphoglycerate kinase 2
Gene Aliases: dJ417L20.2; HEL-S-272; Pgk-2; PGK2; PGKB; PGKPS; Tcp-2
UniProt ID: (Human) P07205, (Pig) Q6RI85, (Mouse) P09041
Entrez Gene ID: (Human) 5232, (Pig) 396673, (Rat) 316265, (Mouse) 18663
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support