Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MAAPEGSGLG EDARLDQETA QWLRWDKNSL TLEAVKRLIA EGNKEELRKC FGARMEFGTA GLRAAMGPGI SRMNDLTIIQ TTQGFCRYLE
Catalyzes the conversion of the nucleoside breakdown products ribose-1-phosphate and deoxyribose-1-phosphate to the corresponding 5-phosphopentoses. May also catalyze the interconversion of glucose-1-phosphate and glucose-6-phosphate. Has low glucose 1,6-bisphosphate synthase activity. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glucose phosphomutase 2; PGM 2; Phosphodeoxyribomutase; Phosphoglucomutase-2; Phosphopentomutase
Gene Aliases: MSTP006; PGM2
UniProt ID: (Human) Q96G03
Entrez Gene ID: (Human) 55276
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support