Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
PGM2L1 is the Glucose 1, 6-bisphosphate synthase using 1, 3-bisphosphoglycerate as a phosphate donor and a series of 1-phosphate sugars as acceptors, including glucose 1-phosphate, mannose 1-phosphate, ribose 1-phosphate and deoxyribose 1-phosphate. 5 or 6-phosphosugars are bad substrates, with the exception of glucose 6-phosphate. PGM2L1 also synthesizes ribose 1, 5-bisphosphate. PGM2L1 has only low phosphopentomutase and phosphoglucomutase activities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ32029; Glucose 1,6-bisphosphate synthase; phosphoglucomutase 2-like 1; Phosphoglucomutase-2-like 1; PMMLP
Gene Aliases: BM32A; PGM2L1; PMMLP
UniProt ID: (Human) Q6PCE3
Entrez Gene ID: (Human) 283209, (Rat) 685076
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support