Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalyzes the conversion of GlcNAc-6-P into GlcNAc-1-P during the synthesis of uridine diphosphate/UDP-GlcNAc, a sugar nucleotide critical to multiple glycosylation pathways including protein N- and O-glycosylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acetylglucosamine phosphomutase; GlcNAc-P mutase; N-acetylglucosamine-phosphate mutase; N-acetylglucosamine-phosphate mutase 1; PAGM; PGM 3; Phosphoacetylglucosamine mutase; Phosphoglucomutase-3
Gene Aliases: 2810473H05Rik; AGM1; BB187688; C77933; IMD23; PAGM; PGM 3; Pgm-3; PGM3
UniProt ID: (Human) O95394, (Mouse) Q9CYR6
Entrez Gene ID: (Human) 5238, (Mouse) 109785
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support