Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAAEAGGD DARCVRLSAE RAQALLADVD TLLFDCDGVL WRGETAVPGA PEALRALRAR GKRLGFITNN SSKTRAAYAE KLRRLGFGGP AGPGASLEVF GTAYCTALYL RQRLAGAPAP KAYVLGSPAL AAELEAVGVA SVGVGPEPLQ GEGPGDWLHA PLEPDVRAVV VGFDPHFSYM KLTKALRYLQ QPGCLLVGTN MDNRLPLENG RFIAGTGCLV RAVEMAAQRQ ADIIGKPSRF IFDCVSQEYG INPERTVMVG DRLDTDILLG ATCGLKTILT LTGVSTLGDV KNNQESDCVS KKKMVPDFYV DSIADLLPAL QG (1-321 aa encoded by BC157035)
Glycerol-3-phosphate phosphatase hydrolyzing glycerol-3-phosphate into glycerol. Thereby, regulates the cellular levels of glycerol-3-phosphate a metabolic intermediate of glucose, lipid and energy metabolism. Was also shown to have a 2-phosphoglycolate phosphatase activity and a tyrosine-protein phosphatase activity. However, their physiological relevance is unclear. In vitro, has also a phosphatase activity toward ADP, ATP, GDP and GTP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aspartate-based ubiquitous Mg(2+)-dependent phosphatase; AUM; G3PP; Glycerol-3-phosphate phosphatase; PGP; PGPase; Phosphoglycolate phosphatase
Gene Aliases: 1700012G19Rik; AI481330; AUM; G3PP; PGP; PGPase
UniProt ID: (Human) A6NDG6, (Mouse) Q8CHP8
Entrez Gene ID: (Human) 283871, (Mouse) 67078
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support