Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In Western blots, no cross-reactivity with recombinant mouse (rm) PGRP-S and rmPGRP-1 beta is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
Peptidoglycan recognition proteins, such as PGRPL, are part of the innate immune system and recognize peptidoglycan, a ubiquitous component of bacterial cell walls.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N-acetylmuramoyl-L-alanine amidase; Peptidoglycan recognition protein 2; Peptidoglycan recognition protein long; PGRP-L; TagL
Gene Aliases: C730002N09Rik; Pglyrp2; Pglyrpl; PGRP-L; Pgrpl; tagL
UniProt ID: (Mouse) Q8VCS0
Entrez Gene ID: (Mouse) 57757
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support