Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: RESVFTVEGG HRAIFFNRIG GVQQDTILAE GLHFRIPWFQ YPIIYDIRAR PRKISSPTGS KDLQMVNISL RVLSRPNAQE LPSMYQRLGL DYEERVLPSI VNEVLKSVVA KFNASQLITQ RAQVSLLIRR ELTERAKDFS LILDDVAITE LSFSREYTAA VEAKQVAQQE AQRAQFLVEK AKQEQRQKIV QAEGEAEAAK MLGEALSKNP GYIKLRKIRA AQNISKTIAT SQNRIYLTAD NLVLNLQDES FTRGSDSLIK GKK
PHB2 acts as a mediator of transcriptional repression by nuclear hormone receptors via recruitment of histone deacetylases. This protein functions as an estrogen receptor (ER)-selective coregulator that potentiates the inhibitory activities of antiestrogens and represses the activity of estrogens. It competes with NCOA1 for modulation of ER transcriptional activity. It is probably involved in regulating mitochondrial respiration activity and in aging.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell associated protein; B-cell receptor-associated protein BAP37; D-prohibitin; MGC117268; PHB2; Prohibitin-2; Repressor of estrogen receptor activity
Gene Aliases: BAP; Bap37; BCAP37; hBAP; p22; PHB2; PNAS-141; REA
UniProt ID: (Human) Q92978
Entrez Gene ID: (Human) 11331
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support