Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLQEQVSEY LGVTSFKRKY PERRDLSHKE KLYLRELNVI TETQCTLGLT ALRSDEVIDL MIKEYPAKHA EYSVILQEKE RQRITDHYKE YSQMQQQNTQ KVEASKVPEY IKKAAKKAAE FNSNLNRERM EERRAYFDLQ THVIQVPQGK YKVLPTERTK VSSYPVALIP GQFQEYYKRY SPDELRYLPL NTALYEPPLD PELPALDSDG DSDDGEDGRG DEKRKNKGTS DSSSGNVSEG ESPPDSQEDS FQGRQKSKDK AATPRKDGPK RSVLSKSVPG YKPKVIPNAI CGICLKGKES NKKGKAESLI HCSQCENSGH PSCLDMTMEL VSMIKTYPWQ CMECKTCIIC GQPHHEEEMM FCDMCDRGYH TFCVGLGAIP SGRWICDCCQ RAPPTPRKVG RRGKNSKEG (1-408 aa encoded by BC020954 )
This gene contains a predicted ORF that encodes a protein with two zinc finger domains. The function of the encoded protein is not known. Sequence analysis suggests that multiple alternatively spliced transcript variants are derived from this gene but the full-length nature of only two of them is known. These two splice variants encode different isoforms. A pseudogene for this gene is located on Xq28.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BAF45a; BRG1-associated factor 45a; BRG1-associated factor, 45-KD, A; PHD finger protein 10; PHD finger protein 10, isoform a; PHD zinc finger protein XAP135; XAP135
Gene Aliases: 1810055P05Rik; AV024533; BAF45A; PHF10; XAP135
UniProt ID: (Human) Q8WUB8, (Mouse) Q9D8M7
Entrez Gene ID: (Human) 55274, (Mouse) 72057
Molecular Function:
histone modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support