Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
The recommended shelf life for this product is 1 year from date of receipt.
Acts as a negative regulator of autophagy, through promoting ubiquitination and degradation of LRSAM1, an E3 ubiquitin ligase that promotes autophagy in response to starvation or infecting bacteria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ16355; MGC2941; PDH-containing protein JUNE-1; PHD finger protein 23; PHD finger protein 23a
Gene Aliases: hJUNE-1b; PHF23
UniProt ID: (Human) Q9BUL5
Entrez Gene ID: (Human) 79142
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support