Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SFGVQQLPLE EIWPLCDFIT VHTPLLPSTT GLLNDNTFAQ CKKGVRVVNC ARGGIVDEGA LLRALQSGQC AGAALDVFTE EPPRDRALVD HENVISCPHL GASTKEAQSR CGEEIAVQFV DMVKGKSLTG VVNAQALTSA FSPHTKPWIG LAEALGTLMR AWAGSPKGTI QVITQGTSLK NAGNCLSPAV IVGLLKEASK QADVNLVNAK LLVKEAGLNV TTSHSPAAPG EQGFGECLLA VALAGAPYQA VGLVQGTTPV LQGLNGAVFR PEVPLRRDLP LLLFRTQTSD PAMLPTMIGL LAEAGVRLLS YQTSLVSDGE TWHVMGISSL LPSLEAWKQH VTEAFQFHF
PHGDH gene encodes the enzyme which is involved in the early steps of L-serine synthesis in animal cells. L-serine is required for D-serine and other amino acid synthesis. The enzyme requires NAD/NADH as a cofactor and forms homotetramers for activity. Mutations in this gene have been found in a family with congenital microcephaly, psychomotor retardation and other symptoms. Multiple alternatively spliced transcript variants have been found, however the full-length nature of most are not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-oxoglutarate reductase; 3-PGDH; 3-phosphoglycerate dehydrogenase; 3-phosphoglycerate dehyrogenase; A10; D-3-phosphoglycerate dehydrogenase; epididymis secretory protein Li 113; Malate dehydrogenase
Gene Aliases: 3-PGDH; 3PGDH; 4930479N23; A10; HEL-S-113; NLS; NLS1; PDG; PGAD; PGD; PGDH; PGDH3; PHGDH; PHGDHD; SERA
UniProt ID: (Human) O43175, (Mouse) Q61753, (Rat) O08651
Entrez Gene ID: (Human) 26227, (Mouse) 236539, (Rat) 58835
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support