Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody (12858-1-AP) detects two bands: 36 kDa and 70 kDa. The 70 kDa band may probably be the dimeric form of PHYH.
Immunogen sequence: MEQLRAAAR LQIVLGHLGR PSAGAVVAHP TSGTISSASF HPQQFQYTLD NNVLTLEQRK FYEENGFLVI KNLVPDADIQ RFRNEFEKIC RKEVKPLGLT VMRDVTISKS EYAPSEKMIT KVQDFQEDKE LFRYCTLPEI LKYVECFTGP NIMAMHTMLI NKPPDSGKKT SRHPLHQDLH YFPFRPSDLI VCAWTAMEHI SRNNGCLVVL PGTHKGSLKP HDYPKWEGGV NKMFHGIQDY EENKARVHLV MEKGDTVFFH PLLIHGSGQN KTQGFRKAIS CHFASADCHY IDVKGTSQEN IEKEVVGIAH KFFGAENSVN LKDIWMFRAR LVKGERTNL (1-338 aa encoded by BC029512 )
PHYH2 catalyzes a carbon-carbon cleavage reaction; cleaves a 2-hydroxy-3-methylacyl-CoA into formyl-CoA and a 2-methyl-branched fatty aldehyde.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1600020H07Rik; HPCL; HPCL2; Lupus nephritis-associated peptide 1; OTTHUMP00000019131; OTTHUMP00000019132; OTTHUMP00000179083; OTTHUMP00000216226; PhyH; PHYH2; Phytanic acid oxidase; phytanoil-CoA alpha hydroxylase; phytanoyl-CoA 2 oxoglutarate dioxygenase; Phytanoyl-CoA alpha-hydroxylase; Phytanoyl-CoA dioxygenase, peroxisomal; phytanoyl-CoA hydroxylase (Refsum disease)
Gene Aliases: AI256161; AI265699; LN1; LNAP1; PAHX; PHYH; PHYH1; RD
UniProt ID: (Human) O14832, (Mouse) O35386, (Rat) P57093
Entrez Gene ID: (Human) 5264, (Mouse) 16922, (Rat) 114209
Molecular Function:
hydroxylase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support