Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
PIAS1 functions as an E3-type small ubiquitin-like modifier (SUMO) ligase, stabilizing the interaction between UBE2I and the substrate, and as a SUMO-tethering factor. This protein plays a crucial role in transcriptional coregulation of various cellular pathways, including the STAT pathway, the p53 pathway and the steroid hormone signaling pathway. It functions in testis as a nuclear receptor transcriptional coregulator and may have a role in androgen receptor initiation and maintenance of spermatogenesis. The effects of transcriptional coregulation, transactivation or silencing, may vary depending upon the biological context.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AR interacting protein; DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1; DEAD/H box-binding protein 1; E3 SUMO-protein ligase PIAS1; E3 SUMO-protein transferase PIAS1; GBP; Gu binding protein; Gu-binding protein; Protein inhibitor of activated STAT protein 1; protein inhibitor of activated STAT-1; RING-type E3 ubiquitin transferase PIAS1; RNA helicase II-binding protein; zinc finger, MIZ-type containing 3
Gene Aliases: 2900068C24Rik; DDXBP1; GBP; GU/RH-II; PIAS1; ZMIZ3
UniProt ID: (Human) O75925, (Mouse) O88907
Entrez Gene ID: (Human) 8554, (Mouse) 56469
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support