Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PNIAVKVNH SYCSVPGYYP SNKPGVEPKR PCRPINLTHL MYLSSATNRI TVTWGNYGKS YSVALYLVRQ LTSSELLQRL KTIGVKHPEL CKALVKEKLR LDPDSEIATT GVRVSLICPL VKMRLSVPCR AETCAHLQCF DAVFYLQMNE KKPTWMCPVC DKPAPYDQLI IDGLLSKILS ECEDADEIEY LVDGSWCPIR AEKERSCSPQ GAILVLGPSD ANGLLPAPSV NGSGALGSTG GGGPVGSMEN GKPGADVVDL TLDSSSSSED EEEEEEEEED EDEEGPRPKR RCPFQKGLVP AC (210-510 aa encoded by BC029874)
Functions as an E3-type small ubiquitin-like modifier (SUMO) ligase, stabilizing the interaction between UBE2I and the substrate, and as a SUMO-tethering factor. Plays a crucial role as a transcriptional coregulation in various cellular pathways, including the STAT pathway, the p53/TP53 pathway, the Wnt pathway and the steroid hormone signaling pathway. Involved in gene silencing. Mediates sumoylation of CEBPA, PARK7, HERC2, MYB, TCF4 and RNF168. In Wnt signaling, represses LEF1 and enhances TCF4 transcriptional activities through promoting their sumoylations. Enhances the sumoylation of MTA1 and may participate in its paralog-selective sumoylation. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: e3 sumo- ligase pias4; E3 SUMO-protein ligase PIAS4; FLJ12419; MGC35296; PIAS-gamma; PIASy; protein inhibitor of activated STAT PIASy; Protein inhibitor of activated STAT protein 4; Protein inhibitor of activated STAT protein gamma; protein inhibitor of activated STAT protein PIASy; protein inhibitor of activated STAT, 4; zinc finger, MIZ-type containing 6
Gene Aliases: PIAS-gamma; PIAS4; PIASG; PIASY; ZMIZ6
UniProt ID: (Human) Q8N2W9, (Mouse) Q9JM05
Entrez Gene ID: (Human) 51588, (Rat) 362827, (Mouse) 59004
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support