Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSGQSLTDRI TAAQHSVTGS AVSKTVCKAT THEIMGPKKK HLDYLIQCTN EMNVNIPQLA DSLFERTTNS SWVVVFKSLI TTHHLMVYGN ERFIQYLASR NTLFNLSNFL DKSGLQGYDM STFIRRYSRY LNEKAVSYRQ VAFDFTKVKR GADGVMRTMN TEKLLKTVPI IQNQMDALLD FNVNSNELTN GVINAAFMLL FKDAIRLFAA YNEGIINLLE KYFDMKKNQC KEGLDIYKKF LTRMTRISEF LKVAEQVGID RGDIPDLSQA PSSLLDALEQ HLASLEGKKI KDSTAASRAT TLSNAVSSLA STGLSLTKVD EREKQAALEE EQARLKALKE QRLKELAKKP HTSLTT
PICALM, also known as the clathrin assembly lymphoid myeloid leukemia (CALM) protein, co-localizes with clathrin at clathrin-coated areas of the plasma membrane and in the Golgi area, and is thought to be an important component of the coated pit internalization machinery. Fusion proteins involving PICALM have been implicated in acute myeloid and T-cell acute lymphoblastic leukemia. PICALM is also expressed in neurons and is present at the synapse where it facilitates the endocytosis of the synaptic vesicle protein VAMP2. Recent studies have identified variants of PICALM associated with Alzheimer's disease and have suggested this protein may play a role in the endocytosis and recycling of the amyloid protein precursor (APP).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CALM; Clathrin assembly lymphoid myeloid leukemia; Clathrin assembly lymphoid myeloid leukemia protein; clathrin-assembly lymphoid leukemia protein; clathrin-assembly lymphoid myeloid leukemia protein; Phosphatidylinositol-binding clathrin assembly protein; rCALM
Gene Aliases: CALM; CLTH; fit-1; Fit1; LAP; mKIAA4114; PAP180; PICALM
UniProt ID: (Human) Q13492, (Mouse) Q7M6Y3, (Rat) O55012
Entrez Gene ID: (Human) 8301, (Mouse) 233489, (Rat) 89816
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support