Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MACRGGAGN GHRASATLSR VSPGSLYTCR TRTHNICMVS DFFYPNMGGV ESHIYQLSQC LIERGHKVII VTHAYGNRKG IRYLTSGLKV YYLPLKVMYN QSTATTLFHS LPLLRYIFVR ERVTIIHSHS SFSAMAHDAL FHAKTMGLQT VFTDHSLFGF ADVSSVLTNK LLTVSLCDTN HIICVSYTSK ENTVLRAALN PEIVSVIPNA VDPTDFTPDP FRRHDSITIV VVSRLVYRKG IDLLSGIIPE LCQKYPDLNF IIGGEGPKRI ILEEVRERYQ LHDRVRLLGA LEHKDVRNVL VQGHIFLNTS LTEAFCMAIV EAASCGLQVV STRVGGIPEV LPENLIILCE PSVKSLCEGL EKAIFQLKSG TLPAPENIHN IVKTFYTWRN VAERTEKVYD RVSVEAVLPM DKRLDRLISH CG (1-421 aa encoded by BC038236)
This gene encodes a protein required for synthesis of N-acetylglucosaminyl phosphatidylinositol (GlcNAc-PI), the first intermediate in the biosynthetic pathway of GPI anchor. The GPI anchor is a glycolipid found on many blood cells and which serves to anchor proteins to the cell surface. Paroxysmal nocturnal hemoglobinuria, an acquired hematologic disorder, has been shown to result from mutations in this gene. Alternate splice variants have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: class A GlcNAc-inositol phospholipid assembly protein; GLCNAC-PI synthesis protein; GPI anchor biosynthesis; N-acetylglucosaminyl-phosphatidylinositol biosynthetic protein; phosphatidylinositol glycan anchor biosynthesis, class A; phosphatidylinositol glycan, class A; Phosphatidylinositol N-acetylglucosaminyltransferase subunit A; Phosphatidylinositol-glycan biosynthesis class A protein; phosphatidylinositol-glycan biosynthesis, class A protein; phosphatidylinositolglycan class A; PIG-A
Gene Aliases: AI194334; GPI3; MCAHS2; PIG-A; PIGA; PNH1
UniProt ID: (Human) P37287, (Mouse) Q64323
Entrez Gene ID: (Human) 5277, (Rat) 363464, (Mouse) 18700
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support